Thermo Scientific Chemicals

4-Chlorobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog number: A15154.22
100 g, Each
Thermo Scientific Chemicals

4-Chlorobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog number: A15154.22
100 g, Each
Quantity
Catalog number: A15154.22
also known as A15154-22
Price (EUR)
Price: 28,00
Online price: 25,20
Your price:
Quantity
-

Chemical Identifiers

CAS
98-56-6
IUPAC Name
1-chloro-4-(trifluoromethyl)benzene
Molecular Formula
C7H4ClF3
InChI Key
QULYNCCPRWKEMF-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(Cl)C=C1
Assay (GC)
≥98.0%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.4445-1.4485 @ 20?C

Description

4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Sensitization
    Category 1
    Skin Sensitization
    Sub-category 1B
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H317 - May cause an allergic skin reaction

    H411 - Toxic to aquatic life with long lasting effects

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    P362 + P364 - Take off contaminated clothing and wash it before reuse