Benzyl 4-chlorophenyl ketone, 98%
Benzyl 4-chlorophenyl ketone, 98%
Benzyl 4-chlorophenyl ketone, 98%
Benzyl 4-chlorophenyl ketone, 98%
Thermo Scientific Chemicals

Benzyl 4-chlorophenyl ketone, 98%

CAS: 1889-71-0 | C14H11ClO | 230.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15959.22
also known as A15959-22
Price (EUR)
580,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
580,00
Each
Add to cart
Chemical Identifiers
CAS1889-71-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
Assay (GC)≥97.5%
Melting Point (clear melt)100.0-107.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only