L-(+)-Rhamnose monohydrate, 98+%
L-(+)-Rhamnose monohydrate, 98+%
L-(+)-Rhamnose monohydrate, 98+%
L-(+)-Rhamnose monohydrate, 98+%
Thermo Scientific Chemicals

L-(+)-Rhamnose monohydrate, 98+%

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16166.14
Price (EUR)
136,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
136,00
Each
Add to cart
Chemical Identifiers
CAS10030-85-0
IUPAC Name(2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
Molecular FormulaC6H12O5
InChI KeySHZGCJCMOBCMKK-HGVZOGFYSA-N
SMILESC[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)5.2-14.13% (0.5-1.5 waters)
Optical Rotation8.0 ± 1.0° (c=10 in water, 20hrs)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
View more
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

Solubility
Soluble in water and methanol. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hausoul, P. J.; Negahdar, L.; Schute, K.; Palkovits, R. A Unravelling the Ru-Catalyzed Hydrogenolysis of Biomass-Based Polyols under Neutral and Acidic Conditions. ChemSusChem 2015, 8 (19), 3323-3330.
  2. Fang, J.; Qin, G.; Ma, J.; She, Y. M. Quantification of plant cell wall monosaccharides by reversed-phase liquid chromatography with 2-aminobenzamide pre-column derivatization and a non-toxic reducing reagent 2-picoline borane. J. Chromatogr. A 2015, 1414, 122-128.