4-n-Butylbenzenesulfonyl chloride, 97%
4-n-Butylbenzenesulfonyl chloride, 97%
4-n-Butylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-n-Butylbenzenesulfonyl chloride, 97%

CAS: 54997-92-1 | C10H13ClO2S | 232.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16577.14
also known as A16577-14
Price (EUR)
230,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
230,00
Each
Chemical Identifiers
CAS54997-92-1
IUPAC Name4-butylbenzene-1-sulfonyl chloride
Molecular FormulaC10H13ClO2S
InChI KeyOVFZELSNOHIDEF-UHFFFAOYSA-N
SMILESCCCCC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentPurchased in U.K. and U.S.
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0% (U.K. specification)
Assay from Supplier's CofA≥96.0% (GC) (U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only