2,6-Di-tert-butyl-4-methylphenol, 99%
2,6-Di-tert-butyl-4-methylphenol, 99%
2,6-Di-tert-butyl-4-methylphenol, 99%
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylphenol, 99%

CAS: 128-37-0 | C15H24O | 220.356 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16863.30
also known as A16863-30
Price (EUR)
28,80
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
28,80
Each
Chemical Identifiers
CAS128-37-0
IUPAC Name2,6-di-tert-butyl-4-methylphenol
Molecular FormulaC15H24O
InChI KeyNLZUEZXRPGMBCV-UHFFFAOYSA-N
SMILESCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5% (U.S. sourced material)
FormCrystals or crystalline powder
Identification (FTIR)Conforms (non-U.S. sourced)
Appearance (Color)White to pale cream
Assay (GC)≥98.5% (non-U.S. sourced material)
View more
2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.

Solubility
Soluble in methanol, toluene, isopropanol, methyl ethyl ketone, acetone, benzene, petroleum ether and ethanol. Insoluble in water.

Notes
Light-sensitive. Keep the container tightly closed in a cool, dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, bases, brass, copper and copper alloys.
RUO – Research Use Only

General References:

  1. Radical inhibitor and antioxidant, readily soluble in nonpolar media. Finds widespread use in the inhibition of peroxide formation in ether solvents and also as a preservative/ antioxidant for unsaturated oils and fats in the food, synthetic rubber and paint industries.
  2. Source of t-butyl group in aromatic substitution reactions. For reviews of the use of t-butyl as a positional protecting group for aromatic substitution reactions, readily removable with, e.g. AlCl 3 in toluene, see: Org. Prep. Proced. Int., 8, 51 (1976); Synthesis, 921 (1979).
  3. Khabibullina, G. A.; Belyaeva, A. S.; Niyazov, N. A.; Movsum-zade, E. M. Simple and low-cost method for processing of waste from production of 2,6-di-tert -butyl-4-methylphenol to give a high-efficiency stabilizer of rubbers and biodiesel fuel. Russ. J. Appl. Chem. 2014, 87 (11), 1680-1685.
  4. Prishchenko, A. A.; Livantsov, M. V.; Novikova, O. P.; Livantsova, L. I.; Milaeva, E. R. Synthesis of Organophosphorus-Substituted Amides of Carbonic Acids with PCHNC(O) and 2,6-Di-tert-butyl-4-methylphenol Fragments. Heteroat. Chem. 2008, 19 (7), 733-737.