2-Fluorophenyl isocyanate, 98%
2-Fluorophenyl isocyanate, 98%
2-Fluorophenyl isocyanate, 98%
Thermo Scientific Chemicals

2-Fluorophenyl isocyanate, 98%

CAS: 16744-98-2 | C7H4FNO | 137.113 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A17466.03
also known as A17466-03
Price (EUR)
25,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
25,20
Each
Chemical Identifiers
CAS16744-98-2
IUPAC Name1-fluoro-2-isocyanatobenzene
Molecular FormulaC7H4FNO
InChI KeyVZNCSZQPNIEEMN-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5105-1.5145 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only