Thermo Scientific Chemicals

Malonamide, 98%, Thermo Scientific Chemicals

Catalog number: A17939.36
500 g, Each
Thermo Scientific Chemicals

Malonamide, 98%, Thermo Scientific Chemicals

Catalog number: A17939.36
500 g, Each
Quantity
Catalog number: A17939.36
also known as A17939-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
108-13-4
IUPAC Name
propanediamide
Molecular Formula
C3H6N2O2
InChI Key
WRIRWRKPLXCTFD-UHFFFAOYSA-N
SMILES
NC(=O)CC(N)=O
Elemental Analysis
(Nitrogen) : 26.75 - 28.12 %
Appearance (Color)
White to very pale cream
Elemental Analysis
(Carbon) : 34.42 - 36.17 %
Form
Crystals or crystalline powder
Purity (DSC)
>97.5%

Description

Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

Solubility
Soluble in HCl.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text