4-Nitrobenzhydrazide, 98+%
4-Nitrobenzhydrazide, 98+%
4-Nitrobenzhydrazide, 98+%
Thermo Scientific Chemicals

4-Nitrobenzhydrazide, 98+%

CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A18016.09
also known as A18016-09
Price (EUR)
73,40
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
73,40
Each
Chemical Identifiers
CAS636-97-5
IUPAC Name4-nitrobenzohydrazide
Molecular FormulaC7H7N3O3
InChI KeyFKZXYJYTUSGIQE-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only