Pentafluoroaniline, 97%
Pentafluoroaniline, 97%
Pentafluoroaniline, 97%
Pentafluoroaniline, 97%
Thermo Scientific Chemicals

Pentafluoroaniline, 97%

CAS: 771-60-8 | C6H2F5N | 183.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19348.22
also known as A19348-22
Price (EUR)
195,50
Online Exclusive
230,00
Save 34,50 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
195,50
Online Exclusive
230,00
Save 34,50 (15%)
Each
Add to cart
Chemical Identifiers
CAS771-60-8
IUPAC Namepentafluoroaniline
Molecular FormulaC6H2F5N
InChI KeyNOXLGCOSAFGMDV-UHFFFAOYSA-N
SMILESNC1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Assay (GC)≥96.0%
Melting Point (clear melt)31.0-40.0?C
Appearance (Color)White to yellow to orange to brown
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

Solubility
Soluble in toluene, and organic solvents. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Funatomi T, et al. Pentafluorophenylammonium triflate (PFPAT): an efficient, practical, and cost-effective catalyst for esterification, thioesterification, transesterification, and macrolactone formation.Green Chem.2006,8(12), 1022-1027.
  2. Parssinen A, et al. Bis (salicylaldiminato) titanium complexes containing bulky imine substituents: Synthesis, characterization and ethene polymerization studies.Eur. J. Inorg. Chem. 2005,2005(11), 2100-2109.