2,4,4-Trimethyl-1-pentene, 99%
2,4,4-Trimethyl-1-pentene, 99%
2,4,4-Trimethyl-1-pentene, 99%
2,4,4-Trimethyl-1-pentene, 99%
Thermo Scientific Chemicals

2,4,4-Trimethyl-1-pentene, 99%

CAS: 107-39-1 | C8H16 | 112.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20187.22
also known as B20187-22
Price (EUR)
158,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
158,00
Each
Add to cart
Chemical Identifiers
CAS107-39-1
IUPAC Name2,4,4-trimethylpent-1-ene
Molecular FormulaC8H16
InChI KeyFXNDIJDIPNCZQJ-UHFFFAOYSA-N
SMILESCC(=C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Colorless
FormLiquid
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

Solubility
Miscible with acetone. Partially soluble in methanol, diethyl ether, benzene, chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agent.
RUO – Research Use Only

General References:

  1. Geukens, I.; De Vos, D. E. Organic Transformations on Metal Nanoparticles: Controlling Activity, Stability, and Recyclability by Support and Solvent Interactions. Langmuir 2013, 29 (10), 3170-3178.
  2. Klajmon, M.; Rehák, K.; Morávek, P.; Matoušová, M. Binary Liquid-Liquid Equilibria of γ-Valerolactone with Some Hydrocarbons. J. Chem. Eng. Data 2015, 60 (5), 1362-1370.
  3. Ning, H.; Gong, C.; Li, Z.; Li, X. Pressure-Dependent Kinetics of Initial Reactions in Iso-octane Pyrolysis. J. Phys. Chem. A 2015, 119 (18), 4093-4107.