5-Mercapto-1-methyltetrazole, 98%
5-Mercapto-1-methyltetrazole, 98%
5-Mercapto-1-methyltetrazole, 98%
5-Mercapto-1-methyltetrazole, 98%
Thermo Scientific Chemicals

5-Mercapto-1-methyltetrazole, 98%

CAS: 13183-79-4 | C2H4N4S | 116.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20443.14
also known as B20443-14
Price (EUR)
95,20
온라인 행사
112,00
Save 16,80 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
95,20
온라인 행사
112,00
Save 16,80 (15%)
Each
Add to cart
Chemical Identifiers
CAS13183-79-4
IUPAC Name1-methyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Molecular FormulaC2H4N4S
InChI KeyXOHZHMUQBFJTNH-UHFFFAOYSA-N
SMILESCN1NN=NC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Melting Point (clear melt)118.0-128.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only