Search Thermo Fisher Scientific
Danger
H302 - Harmful if swallowed
H318 - Causes serious eye damage
H411 - Toxic to aquatic life with long lasting effects
P264 - Wash face, hands and any exposed skin thoroughly after handling
P273 - Avoid release to the environment
P280 - Wear protective gloves/protective clothing/eye protection/face protection
P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting
P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
P310 - Immediately call a POISON CENTER or doctor/physician