Ethyl 4-hydroxyphenylacetate, 98%
Ethyl 4-hydroxyphenylacetate, 98%
Ethyl 4-hydroxyphenylacetate, 98%
Thermo Scientific Chemicals

Ethyl 4-hydroxyphenylacetate, 98%

CAS: 17138-28-2 | C10H12O3 | 180.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21832.09
also known as B21832-09
Price (EUR)
62,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
62,00
Each
Chemical Identifiers
CAS17138-28-2
IUPAC Nameethyl 2-(4-hydroxyphenyl)acetate
Molecular FormulaC10H12O3
InChI KeyHYUPPKVFCGIMDB-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid or liquid as melt
Melting Point (clear melt)33.0-41.5°C
Assay (GC)≥97.5%
Appearance (Color)White to pale cream or clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only