D-(+)-Melezitose hydrate, 99%
D-(+)-Melezitose hydrate, 99%
D-(+)-Melezitose hydrate, 99%
Thermo Scientific Chemicals

D-(+)-Melezitose hydrate, 99%

CAS: 207511-10-2 | C18H32O16 | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B22209.09
also known as B22209-09
Price (EUR)
46,65
線上優惠
52,10
Save 5,45 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
46,65
線上優惠
52,10
Save 5,45 (10%)
Each
Chemical Identifiers
CAS207511-10-2
IUPAC Name(3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyQWIZNVHXZXRPDR-IXUUBQBONA-N
SMILESOC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Loss on Drying≤6.0% (110?C)
Optical Rotation+88 ? 2? (c=4 in water)
FormCrystals or powder or crystalline powder
View more
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Melezitose has been used in a study to assess the identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. It has also been used in a study to investigate new epidemiologic markers of Klebsiella oxytoca strains.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.
RUO – Research Use Only
Maria Silvana Alves et. al.Identification of clinical isolates of indole-positive and indole-negative Klebsiella spp. Journal of Clinical Microbiology. 2006, 44 (10), 3640-3646 .