4,5-Dibromo-o-xylene, 97%
4,5-Dibromo-o-xylene, 97%
4,5-Dibromo-o-xylene, 97%
4,5-Dibromo-o-xylene, 97%
Thermo Scientific Chemicals

4,5-Dibromo-o-xylene, 97%

CAS: 24932-48-7 | C8H8Br2 | 263.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22727.03
also known as B22727-03
Price (EUR)
44,60
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
44,60
Each
Add to cart
Chemical Identifiers
CAS24932-48-7
IUPAC Name1,2-dibromo-4,5-dimethylbenzene
Molecular FormulaC8H8Br2
InChI KeyBCIDDURGCAHERU-UHFFFAOYSA-N
SMILESCC1=CC(Br)=C(Br)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)82.0-92.0?C
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only