2-Chloro-5-nitrobenzonitrile, 99%
2-Chloro-5-nitrobenzonitrile, 99%
2-Chloro-5-nitrobenzonitrile, 99%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzonitrile, 99%

CAS: 16588-02-6 | C7H3ClN2O2 | 182.563 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24287.22
also known as B24287-22
Price (EUR)
205,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
205,00
Each
Chemical Identifiers
CAS16588-02-6
IUPAC Name2-chloro-5-nitrobenzonitrile
Molecular FormulaC7H3ClN2O2
InChI KeyZGILLTVEEBNDOB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (GC)≥98.5%
Melting Point103.5-109.5?C (may be hazy melt)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only