1-Benzyloxy-4-bromobenzene, 97%
1-Benzyloxy-4-bromobenzene, 97%
1-Benzyloxy-4-bromobenzene, 97%
Thermo Scientific Chemicals

1-Benzyloxy-4-bromobenzene, 97%

CAS: 6793-92-6 | C13H11BrO | 263.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24923.03
also known as B24923-03
Price (EUR)
20,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
20,70
Each
Chemical Identifiers
CAS6793-92-6
IUPAC Name1-(benzyloxy)-4-bromobenzene
Molecular FormulaC13H11BrO
InChI KeyOUQSGILAXUXMGI-UHFFFAOYSA-N
SMILESBrC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)57.0-67.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only