2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals
2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals
2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H26135.03
also known as H26135-03
1 g
Catalog number H26135.03
also known as H26135-03
Price (EUR)
177,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
177,00
Each
Chemical Identifiers
CAS146539-83-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)83.0-89.0?C
FormCrystals or powder or crystalline powder
Free acid (titration)≤1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only