Thermo Scientific Chemicals

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), Thermo Scientific Chemicals

Catalog number: H26891.03
1 g, Each
Thermo Scientific Chemicals

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), Thermo Scientific Chemicals

Catalog number: H26891.03
1 g, Each
Quantity
Catalog number: H26891.03
also known as H26891-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
12289-94-0
IUPAC Name
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
Molecular Formula
C16H26Ru
SMILES
[Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol)
319.45
Appearance (Color)
Grey to cream to dark brown
Proton NMR
Conforms to structure
Form
Powder

Description

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures which, in contact with water, emit flammable gases
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    Physical hazards

    H261 - In contact with water releases flammable gases

    Precautionary Statements

    P231 + P232 - Handle and store contents under inert gas. Protect from moisture

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention

    P402 + P404 - Store in a dry place. Store in a closed container