Ethyl 3-cyano-3-phenylpyruvate, 97%
Ethyl 3-cyano-3-phenylpyruvate, 97%
Ethyl 3-cyano-3-phenylpyruvate, 97%
Thermo Scientific Chemicals

Ethyl 3-cyano-3-phenylpyruvate, 97%

CAS: 6362-63-6 | C12H11NO3 | 217.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H26951.06
also known as H26951-06
Price (EUR)
40,80
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
40,80
Each
Chemical Identifiers
CAS6362-63-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Melting Point (clear melt)125.0-131.0°C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only