2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
2,4-Dimethoxybenzylamine, 98%
Thermo Scientific Chemicals

2,4-Dimethoxybenzylamine, 98%

CAS: 20781-20-8 | C9H13NO2 | 167.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27140.06
also known as H27140-06
Price (EUR)
54,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
54,60
Each
Chemical Identifiers
CAS20781-20-8
IUPAC Name1-(2,4-dimethoxyphenyl)methanamine
Molecular FormulaC9H13NO2
InChI KeyQOWBXWFYRXSBAS-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5470-1.5510 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only