4-Bromo-3-chlorobenzoic acid, 97%
4-Bromo-3-chlorobenzoic acid, 97%
4-Bromo-3-chlorobenzoic acid, 97%
4-Bromo-3-chlorobenzoic acid, 97%
Thermo Scientific Chemicals

4-Bromo-3-chlorobenzoic acid, 97%

CAS: 25118-59-6 | C7H4BrClO2 | 235.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27252.14
also known as H27252-14
Price (EUR)
238,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
238,00
Each
Add to cart
Chemical Identifiers
CAS25118-59-6
IUPAC Name4-bromo-3-chlorobenzoic acid
Molecular FormulaC7H4BrClO2
InChI KeyPSKJIHDVFDVNBU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Melting Point (clear melt)220.0-226.0?C
Assay (Silylated GC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only