3-Cyanopyridine-4-boronic acid pinacol ester, 95%
3-Cyanopyridine-4-boronic acid pinacol ester, 95%
3-Cyanopyridine-4-boronic acid pinacol ester, 95%
Thermo Scientific Chemicals

3-Cyanopyridine-4-boronic acid pinacol ester, 95%

CAS: 878194-92-4 | C12H15BN2O2 | 230.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27814.03
also known as H27814-03
Price (EUR)
353,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
353,00
Each
Chemical Identifiers
CAS878194-92-4
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Molecular FormulaC12H15BN2O2
InChI KeyLUDBQHAQFBWGNI-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(C=NC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Proton NMRConforms to structure
Assay (GC)>88.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only