Carbon black, Super P∣r Conductive, 99+% (metals basis)
Carbon black, Super P∣r Conductive, 99+% (metals basis)
Carbon black, Super P∣r Conductive, 99+% (metals basis)
Carbon black, Super P∣r Conductive, 99+% (metals basis)
Thermo Scientific Chemicals

Carbon black, Super P∣r Conductive, 99+% (metals basis)

Carbon black, Super P, >-99%, CAS Number-1333-86-4, 25g, (metals basis), Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S., 215-609-9, 12.01, H351, GHS H Statement H228-H351 Flammable solid. Suspected of causing cancer. | CAS: 1333-86-4 | C | 12.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H30253.14
Price (EUR)
166,65
Online Exclusive
185,00
Save 18,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
166,65
Online Exclusive
185,00
Save 18,35 (10%)
Each
Chemical Identifiers
CAS1333-86-4
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
CommentAbsorption Stiffness Value: 30.0-34.0 ml/5g
CommentDensity: 160±20 kg/m³
CommentMoisture Content (As packed): ≤0.3%
View more
Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Verónica Palomaresa,; Aintzane Goñia,; Izaskun Gil de Muroa,; Iratxe de Meatzab,l Miguel Bengoecheab,; Igor Canteroc,; Teófilo Rojo. Conductive additive content balance in Li-ion battery cathodes: Commercial carbon blacks vs. in situ carbon from LiFePO4/C composites. Journal name. 2010, 195 (22),7661-7668.
  2. RM. Gnanamuthu,; Chang Woo Lee. Electrochemical properties of Super P carbon black as an anode active material for lithium-ion batteries. Materials Chemistry and Physics. 2011, 103 (3),831-834.