3-Chloro-2-methylbenzeneboronic acid, 97%
3-Chloro-2-methylbenzeneboronic acid, 97%
3-Chloro-2-methylbenzeneboronic acid, 97%
Thermo Scientific Chemicals

3-Chloro-2-methylbenzeneboronic acid, 97%

CAS: 313545-20-9 | C7H8BClO2 | 170.399 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H30893.03
also known as H30893-03
Price (EUR)
164,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
164,00
Each
Chemical Identifiers
CAS313545-20-9
IUPAC Name(3-chloro-2-methylphenyl)boronic acid
Molecular FormulaC7H8BClO2
InChI KeyBJPNVVXTUYMJPN-UHFFFAOYSA-N
SMILESCC1=C(C=CC=C1Cl)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only