4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%
Thermo Scientific Chemicals

4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%

CAS: 104460-70-0 | C7H4BrF4N | 258.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H31727.03
also known as H31727-03
Price (EUR)
20,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
20,00
Each
Chemical Identifiers
CAS104460-70-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown or purple
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)31.0-41.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline has antimicrobial effect.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. G. J. Fox,; G. Hallas,; J. D. Hepworth,; K. N. Paskins. para-Bromination of Aromatic Amines: 4-Bromo-N,N-dimethyl-3-(trifluoromethyl)aniline. Organic Syntheses.
  2. Bhawani Singh Rathore,; M. Kumar. Synthesis of 7-chloro-5-trifluoromethyl/7-fluoro/7-trifluoromethyl-4H-1,4-benzothiazines as antimicrobial agents. Bioorganic & Medicinal Chemistry. 2006, 14 (16), 5678-5682.