2,6-Difluoro-4-hydroxybenzaldehyde, 95%
2,6-Difluoro-4-hydroxybenzaldehyde, 95%
2,6-Difluoro-4-hydroxybenzaldehyde, 95%
Thermo Scientific Chemicals

2,6-Difluoro-4-hydroxybenzaldehyde, 95%

CAS: 532967-21-8 | C7H4F2O2 | 158.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H31974.03
also known as H31974-03
Price (EUR)
83,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
83,40
Each
Chemical Identifiers
CAS532967-21-8
IUPAC Name2,6-difluoro-4-hydroxybenzaldehyde
Molecular FormulaC7H4F2O2
InChI KeyROAQMGJHSNIROA-UHFFFAOYSA-N
SMILESOC1=CC(F)=C(C=O)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)174.0-187.0?C
Appearance (Color)Cream to brown
Assay (GC)≥94.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only