3-Bromofuro[3,2-b]pyridine, 96%
3-Bromofuro[3,2-b]pyridine, 96%
3-Bromofuro[3,2-b]pyridine, 96%
3-Bromofuro[3,2-b]pyridine, 96%
Thermo Scientific Chemicals

3-Bromofuro[3,2-b]pyridine, 96%

CAS: 92404-63-2 | C7H4BrNO | 198.019 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33652.03
also known as H33652-03
Price (EUR)
195,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
195,00
Each
Add to cart
Chemical Identifiers
CAS92404-63-2
IUPAC Name3-bromofuro[3,2-b]pyridine
Molecular FormulaC7H4BrNO
InChI KeyKHMAAFDMZSYVEF-UHFFFAOYSA-N
SMILESBrC1=COC2=CC=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
Melting Point (clear melt)49.0-58.0?C
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
Morita, H.; Shiotani, S. Furopyridines. VI. Preparation and reactions of 2- and 3-substituted furo[2,3-b]pyridines. Tetrahedron Lett. 1986, 23 (5), 1465-1469.