11-Aminoundecyltriethoxysilane, 90+%
11-Aminoundecyltriethoxysilane, 90+%
11-Aminoundecyltriethoxysilane, 90+%
Thermo Scientific Chemicals

11-Aminoundecyltriethoxysilane, 90+%

CAS: 116821-45-5 | C17H39NO3Si | 333.588 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33835.03
also known as H33835-03
Price (EUR)
475,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
475,00
Each
Chemical Identifiers
CAS116821-45-5
IUPAC Name(11-aminoundecyl)triethoxysilane
Molecular FormulaC17H39NO3Si
InChI KeyLPWZCJFZJCOBHO-UHFFFAOYSA-N
SMILESCCO[Si](CCCCCCCCCCCN)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥90.0%
Identification (FTIR)Conforms
Refractive Index1.4340-1.4420 @ 20°C
Appearance (Color)Clear colorless to pale yellow
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Da-Ren Chiou; Kuan-Yu Yeh; and Li-Jen Chen. Adjustable pretilt angle of nematic 4-n-pentyl-4'-cyanobiphenyl on self-assembled monolayers formed from organosilanes on square-wave grating silica surfaces. Appl. Phys. Lett. 200688 (13), 133123
  2. Max Schütz; Mohammad Salehi; and Sebastian Schlücker.Direct Silica Encapsulation of Self-Assembled-Monolayer-Based Surface-Enhanced Raman Scattering Labels with Complete Surface Coverage of Raman Reporters by Noncovalently Bound Silane Precursors. Chemistry - An Asian Journal. 20149 (8), 2219-2224