Potassium tetrakis(pentafluorophenyl)borate, 97%
Potassium tetrakis(pentafluorophenyl)borate, 97%
Potassium tetrakis(pentafluorophenyl)borate, 97%
Potassium tetrakis(pentafluorophenyl)borate, 97%
Thermo Scientific Chemicals

Potassium tetrakis(pentafluorophenyl)borate, 97%

CAS: 89171-23-3 | C24BF20K | 718.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36527.06
Price (EUR)
419,90
Online Exclusive
494,00
Save 74,10 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
419,90
Online Exclusive
494,00
Save 74,10 (15%)
Each
Add to cart
Chemical Identifiers
CAS89171-23-3
IUPAC Namepotassium tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular FormulaC24BF20K
InChI KeyGYBHRIJOPWTIKA-UHFFFAOYSA-N
SMILES[K+].FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR≥96.0%
Fluorine-19 NMRConforms to structure
FormPowder
Appearance (Color)Pale cream to cream to pale brown
Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hou, B.; Bu, W.; Luo, G.; Vanýsek, P.; Schlossman, M. L. Ion Distributions at Electrified Water-Organic Interfaces: PB-PMF Calculations and Impedance Spectroscopy Measurements. J. Electrochem. Soc. 2015, 162 (12), H890-H897.
  2. Hu, X.; Soleilhavoup, M.; Melaimi, M.; Chu, J.; Bertrand, G. Air-Stable (CAAC) CuCl and (CAAC) CuBH4 Complexes as Catalysts for the Hydrolytic Dehydrogenation of BH3NH3. Angew. Chem. 2015, 54 (20), 6008-6011.