6-Bromo-1,8-naphthyridin-2(1H)-one, 98%
6-Bromo-1,8-naphthyridin-2(1H)-one, 98%
6-Bromo-1,8-naphthyridin-2(1H)-one, 98%
Thermo Scientific Chemicals

6-Bromo-1,8-naphthyridin-2(1H)-one, 98%

CAS: 72754-05-3 | C8H5BrN2O | 225.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50042.03
also known as H50042-03
Price (EUR)
336,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
336,00
Each
Chemical Identifiers
CAS72754-05-3
IUPAC Name6-bromo-1,2-dihydro-1,8-naphthyridin-2-one
Molecular FormulaC8H5BrN2O
InChI KeyBCEWGGGNOQNYKK-UHFFFAOYSA-N
SMILESBrC1=CN=C2NC(=O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
Assay (HPLC)>97.5%
FormPowder
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Valentina Lucchesi; Dow P.Hurst; Derek M.Shore; Simone Bertini; Brandie M.Ehrmann; Marco Allara; Lyle Lawrence; Alessia Ligresti; Filippo Minutolo; Giuseppe Saccomanni; Haleli Sharir; Marco Macchia; Vincenzo Di Marzo; Mary E.Abood; Patricia H.Reggio; Clementina Manera. CB2-Selective cannabinoid receptor ligands: Synthesis, pharmacological evaluation, and molecular modeling investigation of 1,8-naphthyridin-2(1H)-one-3-carboxamides. J. Med. Chem. 2014, 57,(21), 8777-8791.