Thermo Scientific Chemicals

Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: H50153.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog number: H50153.06
5 g, Each
Quantity
Catalog number: H50153.06
also known as H50153-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
902837-56-3
IUPAC Name
ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
Molecular Formula
C9H8ClNO3
InChI Key
ZTIMVJIGNBYGJG-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=O)C1=C(Cl)N=CC=C1
Assay (unspecified)
>96.0%
Form
Liquid
Appearance (Color)
Reddish brown

Description

Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text