(R)-3-Amino-3-phenylpropionic acid, 95%
(R)-3-Amino-3-phenylpropionic acid, 95%
(R)-3-Amino-3-phenylpropionic acid, 95%
Thermo Scientific Chemicals

(R)-3-Amino-3-phenylpropionic acid, 95%

CAS: 83649-48-3 | C9H12ClNO2 | 201.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52034.06
also known as H52034-06
Price (EUR)
837,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
837,00
Each
Chemical Identifiers
CAS83649-48-3
IUPAC Name(3R)-3-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyUJOYFRCOTPUKAK-MRVPVSSYSA-N
SMILESN[C@H](CC(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Supplier's CofA≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only