2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one, 97%
2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one, 97%
2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one, 97%
Thermo Scientific Chemicals

2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one, 97%

CAS: 3158-91-6 | C13H8ClNO2 | 245.662 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54266.03
also known as H54266-03
Price (EUR)
126,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
126,00
Each
Specifications
CAS3158-91-6
Chemical Name or Material2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one
EINECS Number221-602-1
Formula Weight245.66
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only