1,3-Benzodioxole-5-sulfonyl chloride, 95%
1,3-Benzodioxole-5-sulfonyl chloride, 95%
1,3-Benzodioxole-5-sulfonyl chloride, 95%
Thermo Scientific Chemicals

1,3-Benzodioxole-5-sulfonyl chloride, 95%

CAS: 115010-10-1 | C7H5ClO4S | 220.623 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54755.03
also known as H54755-03
Price (EUR)
185,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
185,00
Each
Specifications
CAS115010-10-1
Chemical Name or Material1,3-Benzodioxole-5-sulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight220.63
Health Hazard 1H314-H335
View more
1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ronald G.Sherrill; Eric S.Furfine; Richard J.Hazen; John F.Miller; David J.Reynolds; Douglas M.Sammond; Andrew Spaltenstein; Pat Wheelan; Lois L.Wright. Synthesis and antiviral activities of novel N-alkoxy-arylsulfonamide-based HIV protease inhibitors. Bioorganic & Medicinal Chemistry Letters. 2005, 15,(15), 3560-3564.
  2. Emile J.Velthuisen; Todd M.Baughman; Brian A.Johns; David P.Temelkoff; Jason G.Weatherhead. Synthesis and pharmacokinetic profile of highly deuterated brecanavir analogs. European Journal of Medicinal Chemistry. 2013, 63,202-212.