4-Iodo-2-(trifluoromethyl)benzonitrile, 97%
4-Iodo-2-(trifluoromethyl)benzonitrile, 97%
4-Iodo-2-(trifluoromethyl)benzonitrile, 97%
Thermo Scientific Chemicals

4-Iodo-2-(trifluoromethyl)benzonitrile, 97%

CAS: 101066-87-9 | C8H3F3IN | 297.019 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55609.06
also known as H55609-06
Price (EUR)
134,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
134,00
Each
Chemical Identifiers
CAS101066-87-9
IUPAC Name4-iodo-2-(trifluoromethyl)benzonitrile
Molecular FormulaC8H3F3IN
InChI KeyRBRTXKYFMXVIER-UHFFFAOYSA-N
SMILESFC(F)(F)C1=C(C=CC(I)=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-(trifluoromethyl)benzonitrile is used as pharmaceutical intermediates

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only