Chemical Identifiers
CAS41372-08-1
IUPAC Namebis((2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid) trihydrate
Molecular FormulaC20H32N2O11
InChI KeyYKFCISHFRZHKHY-NGQGLHOPSA-N
SMILESO.O.O.C[C@](N)(CC1=CC=C(O)C(O)=C1)C(O)=O.C[C@](N)(CC1=CC=C(O)C(O)=C1)C(O)=O
View more