Benzyl 2-bromoethyl ether, 97%
Benzyl 2-bromoethyl ether, 97%
Benzyl 2-bromoethyl ether, 97%
Benzyl 2-bromoethyl ether, 97%
Thermo Scientific Chemicals

Benzyl 2-bromoethyl ether, 97%

CAS: 1462-37-9 | C9H11BrO | 215.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H56872.14
also known as H56872-14
Price (EUR)
237,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
237,00
Each
Add to cart
Chemical Identifiers
CAS1462-37-9
IUPAC Name[(2-bromoethoxy)methyl]benzene
Molecular FormulaC9H11BrO
InChI KeyFWOHDAGPWDEWIB-UHFFFAOYSA-N
SMILESBrCCOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)96.0% min
Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
RUO – Research Use Only

General References:

  1. Martin W. M. Fijten.; Claudia Haensch.; Bart M. van Lankvelt.; Richard Hoogenboom.; Ulrich S. Schubert. Clickable Poly(2-Oxazoline)s as Versatile Building Blocks. Macromolecular Chemistry and Physics. 2008, 209 (18),1887-1895.
  2. Britt A. Minch.; Wei Xia.; Carrie L. Donley.; Ryan M. Hernandez.; Chet Carter.; Michael D. Carducci.; Alice Dawson.; David F. O'Brien.; Neal R. Armstrong. Octakis(2-benzyloxyethylsulfanyl) Copper (II) Phthalocyanine: A New Liquid Crystalline Discotic Material with Benzyl-Terminated, Thioether-Linked Side Chains. Chem. Mater. 2005, 17 (7),1618-1627.