Violanthrone-79, 98%
Violanthrone-79, 98%
Violanthrone-79, 98%
Thermo Scientific Chemicals

Violanthrone-79, 98%

CAS: 85652-50-2 | C50H48O4 | 712.93 g/mol
Have Questions?
Catalog NumberQuantity
H59616.03
also known as H59616-03
1 g
Catalog number H59616.03
also known as H59616-03
Price (EUR)
306,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
306,00
Each
Chemical Identifiers
CAS85652-50-2
IUPAC Name30,34-bis(octyloxy)nonacyclo[18.10.2.2²,⁵.0³,¹⁶.0⁴,¹³.0⁶,¹¹.0¹⁷,³¹.0²²,²⁷.0²⁸,³²]tetratriaconta-1(30),2(34),3,5(33),6,8,10,13,15,17,19,22,24,26,28,31-hexadecaene-12,21-dione
Molecular FormulaC50H48O4
InChI KeyLLPQZABTDLOYAL-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=C2C3=C(OCCCCCCCC)C=C4C5=CC=CC=C5C(=O)C5=CC=C(C6=CC=C7C(=O)C8=CC=CC=C8C(=C1)C7=C26)C3=C45
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.

Solubility
Soluble in chloroform.

Notes
Refrigerated. Incompatible with strong oxi
RUO – Research Use Only

General References:

  1. Francisco López-Linares.; Lante Carbognani.; Manuel F. González.; Clementina Sosa-Stull.; Maria Figueras.; Pedro Pereira-Almao. Quinolin-65 and Violanthrone-79 as Model Molecules for the Kinetics of the Adsorption of C7 Athabasca Asphaltene on Macroporous Solid Surfaces. Energy Fuels. 2006, 20 (6),2748-2750.
  2. Fahrettin Yakuphanoglu. Interface control of conventional n-type silicon/metal by n-channel organic semiconductor. Microelectron. Eng. 2010, 87 (10), 1884-1888.