Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%
Thermo Scientific Chemicals

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%

CAS: 811862-77-8 | C42H44O10Pd | 815.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61964.06
also known as H61964-06
Price (EUR)
348,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
348,00
Each
Chemical Identifiers
CAS811862-77-8
IUPAC Namebis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Molecular FormulaC42H44O10Pd
InChI KeyRTGAJOPJZJDWAX-UHFFFAOYSA-N
SMILES[Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>98.0%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Jan Streuff; David E. White; Scott C. Virgil; & Brian M. Stoltz. A palladium-catalysed enolate alkylation cascade for the formation of adjacent quaternary and tertiary stereocentres. Nature Chemistry. 2010, 2 192-196.
  2. Amarajothi Dhakshinamoorthy; Kasi Pitchumani. Clay-anchored non-heme iron-salen complex catalyzed cleavage of Cdouble bond; length as m-dashC bond in aqueous medium. Tetrahedron. 2006, 62(42), 9911-9918.