trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%
trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%
trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%
Thermo Scientific Chemicals

trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

CAS: 209128-50-7 | C12H21NO4 | 243.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H62007.06
also known as H62007-06
Price (EUR)
405,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
405,00
Each
Chemical Identifiers
CAS209128-50-7
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular FormulaC12H21NO4
InChI KeyQJEQJDJFJWWURK-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC1CCCCC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White crystalline
Assay (unspecified)>96.0%
Impurity content≤0.1% cis-Isomer content
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only