N-Boc-3-cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
N-Boc-3-cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
N-Boc-3-cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-3-cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62517.03
also known as H62517-03
1 g
Catalog number H62517.03
also known as H62517-03
Price (EUR)
242,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
242,00
Each
Chemical Identifiers
CAS131980-30-8
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-cyanophenyl)propanoate
Molecular FormulaC15H17N2O4
InChI KeyFDQDHMZKOPOWFE-LBPRGKRZSA-M
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(=C1)C#N)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only