Methyl 3-bromo-5-fluorophenylacetate, 96%
Methyl 3-bromo-5-fluorophenylacetate, 96%
Methyl 3-bromo-5-fluorophenylacetate, 96%
Thermo Scientific Chemicals

Methyl 3-bromo-5-fluorophenylacetate, 96%

CAS: 937732-38-2 | C9H8BrFO2 | 247.063 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62561.06
also known as H62561-06
Price (EUR)
832,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
832,00
Each
Chemical Identifiers
CAS937732-38-2
IUPAC Namemethyl 2-(3-bromo-5-fluorophenyl)acetate
Molecular FormulaC9H8BrFO2
InChI KeyAJSCFDHWZUZCNE-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC(F)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)Oily
Assay by NMR>95.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only