3-Bromo-2,6-difluoroaniline, 96%
3-Bromo-2,6-difluoroaniline, 96%
3-Bromo-2,6-difluoroaniline, 96%
Thermo Scientific Chemicals

3-Bromo-2,6-difluoroaniline, 96%

CAS: 1262198-07-1 | C6H4BrF2N | 208.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62609.03
also known as H62609-03
Price (EUR)
238,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
238,00
Each
Chemical Identifiers
CAS1262198-07-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)≥95.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only