3-(1-Naphthyl)-L-alanine, 95%
3-(1-Naphthyl)-L-alanine, 95%
3-(1-Naphthyl)-L-alanine, 95%
Thermo Scientific Chemicals

3-(1-Naphthyl)-L-alanine, 95%

CAS: 55516-54-6 | C13H13NO2 | 215.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63586.03
also known as H63586-03
Price (EUR)
61,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
61,40
Each
Chemical Identifiers
CAS55516-54-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. M Rodriguez; N Bernad; MC Galas; MF Lignon; J Laur; A Aumelas; J Martinez. Synthèse et activités biologiques d'analogues de la cholécystokinine substitués en position 30 par la 3-(1-naphtyl) l-alanine [Nal(1)] et la 3-(2-naphthyl) l-alanine [Nal(2)]. European Journal of Medicinal Chemistry. 1991, 26(3), 245-253.
  2. Dr. Abhishek Chatterjee; Han Xiao; Dr. Peng-Yu Yang; Gautam Soundararajan; Prof. Dr. Peter G. Schultz. A Tryptophanyl-tRNA Synthetase/tRNA Pair for Unnatural Amino Acid Mutagenesis in E. coli reaction. Angewandte Chemie International. 2013,52(19), 5106-5109.