Thermo Scientific Chemicals

1-Boc-indoline-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H63883.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-indoline-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H63883.03
1 g, Each
Quantity
Catalog number: H63883.03
also known as H63883-03
Price (EUR)
Quantity
-

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Sensitization
    Category 1
    Acute aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H317 - May cause an allergic skin reaction

    H400 - Very toxic to aquatic life

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention