2-Cyano-5-(trifluoromethyl)pyridine, 98%
2-Cyano-5-(trifluoromethyl)pyridine, 98%
2-Cyano-5-(trifluoromethyl)pyridine, 98%
Thermo Scientific Chemicals

2-Cyano-5-(trifluoromethyl)pyridine, 98%

CAS: 95727-86-9 | C7H3F3N2 | 172.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64486.06
also known as H64486-06
Price (EUR)
119,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
119,00
Each
Chemical Identifiers
CAS95727-86-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Ramajeyam Selvaraj; Joseph M. Fox. An efficient and mild oxidant for the synthesis of s-tetrazines. Tetrahedron Letters.2014 55(34), 4795-4797.