5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%
5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%
5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%
Thermo Scientific Chemicals

5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%

CAS: 80194-69-0 | C7H4F3NO2 | 191.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64937.14
also known as H64937-14
Price (EUR)
613,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
613,00
Each
Chemical Identifiers
CAS80194-69-0
IUPAC Name5-(trifluoromethyl)pyridine-2-carboxylic acid
Molecular FormulaC7H4F3NO2
InChI KeyNJHGVAYLDHROPT-UHFFFAOYSA-N
SMILESOC(=O)C1=NC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Roger Crettaz; Jérôme Waser; and Yves Bessard. Synthetic Process Development and Scale Up of Palladium-Catalyzed Alkoxycarbonylation of Chloropyridines. Org. Proc. Res. Dev.2001 5(6), 572-574.