MOPS, 0.5M buffer soln., pH 7.0
MOPS, 0.5M buffer soln., pH 7.0
MOPS, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals

MOPS, 0.5M buffer soln., pH 7.0

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J60328.AK
also known as J60328-AK
Price (EUR)
103,00
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
103,00
Each
Specifications
CAS1132-61-2
ChEBICHEBI:44115
Chemical Name or MaterialMOPS buffer soln.
Health Hazard 1H315-H319-H335
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only