Sodium citrate, 0.5M buffer soln., pH 6.0
Thermo Scientific Chemicals

Sodium citrate, 0.5M buffer soln., pH 6.0

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61815.AP
also known as J61815-AP
Price (EUR)
100,65
Online Exclusive
112,00
Save 11,35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
100,65
Online Exclusive
112,00
Save 11,35 (10%)
Each
Specifications
Chemical Name or MaterialSodium citrate buffer soln.
ColorColorless
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
View more
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

Solubility
Fully miscible in water.

Notes
Incompatible with heat and oxidizing agents.
RUO – Research Use Only

General References:

  1. Mostafa Mesbah; Usha Premachandran; William B. Whitman. Precise Measurement of the G+C Content of Deoxyribonucleic Acid by High-Performance Liquid Chromatography. Int J Syst Evol Microbiol.1989, 39, 159-167.
  2. Tomas Lindahl; Annika Andersson. Rate of chain breakage at apurinic sites in double-stranded deoxyribonucleic acid. Biochemistry.1972, 11 , (19), 3618-3623.